Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate 95%
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate 95%
APOSOR955867-1G
:
Apollo Scientific
:
Methyl (2s)-2-amino-3-(furan-2-yl)propanoate 95%
APOSOR955867-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
169.178 g/mol
:
Ambient
:
146725-85-1
:
C₈H₁₁NO₃
:
MFCD10565764