Zadání objednávky
Czech Republic
ContactUsLinkComponent
Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride 98%
Irritant
Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride 98%
Catalog # APOSOR955865-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 2-(1H-imidazol-4-yl)acetate hydrochloride 98%
Catalog # APOSOR955865-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    1 g
  • Sekvence:
    3047

Specifications

About this item

 :
 :176.601 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:51718-80-0
Formula:C₆H₈N₂O₂·ClH
MDL Number:MFCD08275082