Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl 2-methylpyrimidine-4-carboxylate 95%
  APOSOR955753-1G
 :  
Methyl 2-methylpyrimidine-4-carboxylate 95%
  APOSOR955753-1G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

 :
 :152.151 g/mol
 :Ambient
 :73955-55-2
 :C₇H₈N₂O₂
 :MFCD13174780