
Methyl 2-(2-bromo-4-chlorophenyl)acetate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:115871-49-3
:
Formula:C₉H₈BrClO₂
MDL Number:MFCD06658159
:263.516 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together