Order Entry
Czech Republic
ContactUsLinkComponent
Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate 95%
Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate 95%
Catalog # APOSOR955588-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate 95%
Catalog # APOSOR955588-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.177 g/mol
Storage Temperature:Ambient
CAS Number:1563651-10-4
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD23716020