(S)-(+)-Methyl indoline-2-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:141410-06-2
:
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD12796094
:177.2 g/mol
Storage Temperature:Freezer
UN:3077
ADR:9+EHS,III
Frequently Bought Together