Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(3-Formylphenyl)phenol
4-(3-Formylphenyl)phenol 98%
4-(3-Formylphenyl)phenol 98%
APOSOR954782-1G
:
Apollo Scientific
:
4-(3-Formylphenyl)phenol 98%
APOSOR954782-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
198.217 g/mol
:
Ambient
:
283147-95-5
:
C₁₃H₁₀O₂
:
MFCD04117372