Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Methoxy-phenyl)-ethylamine 95%
1-(2-Methoxy-phenyl)-ethylamine 95%
Catalog # APOSOR953919-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Methoxy-phenyl)-ethylamine 95%
Catalog # APOSOR953919-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.206 g/mol
Storage Temperature:Ambient
CAS Number:40023-74-3
Formula:C₉H₁₃NO
MDL Number:MFCD00593475