1-(3-Methoxy-phenyl)-n1,n1-dimethyl-ethane-1,2-diamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:855715-34-3
:
Formula:C₁₁H₁₈N₂O
MDL Number:MFCD06655099
:194.273 g/mol
Storage Temperature:Ambient
Frequently Bought Together