(S)-2-Pyridylthio cysteamine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106139-15-5
:
Formula:C₇H₁₀N₂S₂·ClH
MDL Number:MFCD01863453
:222.759 g/mol
Melting Pt:124…126 °C
Storage Temperature:Ambient
Frequently Bought Together