Order Entry
Czech Republic
ContactUsLinkComponent
7-Methyl-1,3-dihydroimidazo[4,5-b]pyridin-2-one 95%
7-Methyl-1,3-dihydroimidazo[4,5-b]pyridin-2-one 95%
Catalog # APOSOR953346-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Methyl-1,3-dihydroimidazo[4,5-b]pyridin-2-one 95%
Catalog # APOSOR953346-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.15 g/mol
Storage Temperature:Ambient
CAS Number:518038-75-0
Formula:C₇H₇N₃O
MDL Number:MFCD14585004