Order Entry
Czech Republic
ContactUsLinkComponent
Cyclopropyl methoxylamine hydrochloride 95%
Cyclopropyl methoxylamine hydrochloride 95%
Catalog # APOSOR953045-1G
Supplier:  Apollo Scientific
CAS Number:  
Cyclopropyl methoxylamine hydrochloride 95%
Catalog # APOSOR953045-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :123.581 g/mol
Storage Temperature:Ambient
CAS Number:74124-04-2
Formula:C₄H₉NO·ClH
MDL Number:MFCD08689925