
4-Cyclopropylbenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:167404-32-2
:
Formula:C₉H₉ClO₂S
MDL Number:MFCD09929602
:216.685 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together