
Methyl 4-(1-aminocyclopropyl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1006037-03-1
:
Formula:C₁₁H₁₃NO₂
MDL Number:MFCD11616340
:191.226 g/mol
Melting Pt:50 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together