
4-(2-Ethoxy-2-oxoethoxy)benzeneboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:282116-97-6
:
Formula:C₁₀H₁₃BO₅
MDL Number:MFCD08235079
:224.018 g/mol
Melting Pt:145…147 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together