Order Entry
Czech Republic
ContactUsLinkComponent
5-(1-Hydroxyethyl)-2-methylpyridine 95%
5-(1-Hydroxyethyl)-2-methylpyridine 95%
Catalog # APOSOR946325-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(1-Hydroxyethyl)-2-methylpyridine 95%
Catalog # APOSOR946325-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :137.179 g/mol
Storage Temperature:Ambient
CAS Number:100189-16-0
Formula:C₈H₁₁NO
MDL Number:MFCD06637422