Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,7-Diazaspiro[3.5]nonan-2-one
1,7-Diazaspiro[3.5]nonan-2-one
1,7-Diazaspiro[3.5]nonan-2-one
APOSOR946098-1G
:
Apollo Scientific
:
1,7-Diazaspiro[3.5]nonan-2-one
APOSOR946098-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
140.183 g/mol
:
Ambient
:
1235440-17-1
:
C₇H₁₂N₂O
:
MFCD14705692