Methyl 2-(4-hydroxy-2-methylphenoxy)acetate 95%
Catalog # APOSOR944537-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.2 g/mol
Storage Temperature:Ambient
CAS Number:317319-10-1
Formula:C₁₀H₁₂O₄
MDL Number:MFCD07778439