2-(2-Chloroethoxy)acetonitrile 95%
Catalog # APOSOR944172-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:119.55 g/mol
Storage Temperature:Ambient
CAS Number:31250-08-5
Formula:C₄H₆ClNO
MDL Number:MFCD19159867