7-Chloro-2,3-dihydro-1,8-naphthyridin-4(1H)-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76629-10-2
:
Formula:C₈H₇ClN₂O
MDL Number:MFCD09834137
:182.607 g/mol
Storage Temperature:Ambient
Frequently Bought Together