Order Entry
Czech Republic
ContactUsLinkComponent
(5-Chloro-2-methoxyphenyl)acetonitrile 95%
(5-Chloro-2-methoxyphenyl)acetonitrile 95%
Catalog # APOSOR942910-1G
Supplier:  Apollo Scientific
CAS Number:  
(5-Chloro-2-methoxyphenyl)acetonitrile 95%
Catalog # APOSOR942910-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.619 g/mol
Storage Temperature:Ambient
CAS Number:7048-38-6
Formula:C₉H₈ClNO
MDL Number:MFCD07348599