Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-(2-chloroethoxy)acetate
Methyl 2-(2-chloroethoxy)acetate 95%
Methyl 2-(2-chloroethoxy)acetate 95%
APOSOR942013-1G
:
Apollo Scientific
:
Methyl 2-(2-chloroethoxy)acetate 95%
APOSOR942013-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
152.576 g/mol
:
Ambient
:
83881-47-4
:
C₅H₉ClO₃
:
MFCD09030969