Order Entry
Czech Republic
ContactUsLinkComponent
N-tert-Butyl-1,1-dimethylpropargylamine 95%
N-tert-Butyl-1,1-dimethylpropargylamine 95%
Catalog # APOSOR938982-5G
Supplier:  Apollo Scientific
CAS Number:  
N-tert-Butyl-1,1-dimethylpropargylamine 95%
Catalog # APOSOR938982-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :139.238 g/mol
Storage Temperature:Ambient
CAS Number:1118-17-8
Formula:C₉H₁₇N
MDL Number:MFCD02093662