(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanol 95%
Catalog # APOSOR938981-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.183 g/mol
Storage Temperature:Ambient
CAS Number:18712-39-5
Formula:C₇H₁₂N₂O
MDL Number:MFCD00810197