Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 1-hydroxycyclohexanecarboxylate 95%
Ethyl 1-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR938628-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR938628-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.222 g/mol
Storage Temperature:Ambient
CAS Number:1127-01-1
Formula:C₉H₁₆O₃
MDL Number:MFCD06797659