Order Entry
Czech Republic
ContactUsLinkComponent
 
Pent-4-yn-1-amine hydrochloride 97%
Catalog # APOSOR938293-500MG
Supplier:  Apollo Scientific
CAS Number:  
Pent-4-yn-1-amine hydrochloride 97%
Catalog # APOSOR938293-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :119.593 g/mol
Storage Temperature:Ambient
CAS Number:173987-24-1
Formula:C₅H₉N·ClH
MDL Number:MFCD22836507