Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate 95%
Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate 95%
Catalog # APOSOR936947-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl (E)-2-cyano-3-ethoxy-but-2-enoate 95%
Catalog # APOSOR936947-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :183.204 g/mol
Storage Temperature:Ambient
CAS Number:35260-93-6
Formula:C₉H₁₃NO₃
MDL Number:MFCD01743485