Order Entry
Czech Republic
ContactUsLinkComponent
 
6-Methyl-2-azaspiro[3.3]heptan-6-ol hydrochloride
Catalog # APOSOR935289-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-2-azaspiro[3.3]heptan-6-ol hydrochloride
Catalog # APOSOR935289-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.645 g/mol
Storage Temperature:Ambient
CAS Number:1638765-02-2
Formula:C₇H₁₃NO·ClH
MDL Number:MFCD28501954