5-Bromobenzo[d]isothiazol-3(2H)-one ≥98%
Catalog # APOSOR935139-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:230.082 g/mol
Storage Temperature:Refrigerator
CAS Number:186584-79-2
Formula:C₇H₄BrNOS
MDL Number:MFCD28671026