
(R)-1,3'-Bipyrrolidine dihydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:913702-34-8
:
Formula:C₈H₁₆N₂·2ClH
MDL Number:MFCD27952641
:213.148 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together