1-(Tetrahydro-2-furanylmethyl)-1H-pyrrole-2-carbaldehyde 95%
Catalog # APOSOR932879-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:179.216 g/mol
Storage Temperature:Ambient
CAS Number:932186-29-3
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD09674984