Order Entry
Czech Republic
ContactUsLinkComponent
1-(3-Nitro-4-pyridinyl)-ethanone 95%
1-(3-Nitro-4-pyridinyl)-ethanone 95%
Catalog # APOSOR931499-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Nitro-4-pyridinyl)-ethanone 95%
Catalog # APOSOR931499-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.134 g/mol
Storage Temperature:Ambient
CAS Number:161871-65-4
Formula:C₇H₆N₂O₃
MDL Number:MFCD03411728