Order Entry
Czech Republic
ContactUsLinkComponent
(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol 97%
(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol 97%
Catalog # APOSOR9308-1G
Supplier:  Apollo Scientific
CAS Number:  
(1-Methyl-5-thien-2-yl-1H-pyrazol-3-yl)methanol 97%
Catalog # APOSOR9308-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.254 g/mol
Melting Pt:56…60 °C
Storage Temperature:Ambient
CAS Number:879896-47-6
Formula:C₉H₁₀N₂OS
MDL Number:MFCD08435882