Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Methoxyphenyl)pyrrolidin-2-one
4-(2-Methoxyphenyl)pyrrolidin-2-one 95%
4-(2-Methoxyphenyl)pyrrolidin-2-one 95%
4-(2-Methoxyphenyl)pyrrolidin-2-one 95%
APOSOR930701-1G
:
Apollo Scientific
:
4-(2-Methoxyphenyl)pyrrolidin-2-one 95%
APOSOR930701-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.226 g/mol
:
Ambient
:
1020718-50-6
:
C₁₁H₁₃NO₂
:
MFCD09955050