Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-(Methylamino)phenyl)ethanone 95%
1-(2-(Methylamino)phenyl)ethanone 95%
Catalog # APOSOR929725-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-(Methylamino)phenyl)ethanone 95%
Catalog # APOSOR929725-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.19 g/mol
Storage Temperature:Ambient
CAS Number:1859-75-2
Formula:C₉H₁₁NO
MDL Number:MFCD19159692