Order Entry
Czech Republic
ContactUsLinkComponent
5-(2-Chloroethyl)-1,3-benzodioxole 97%
5-(2-Chloroethyl)-1,3-benzodioxole 97%
Catalog # APOSOR9297-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(2-Chloroethyl)-1,3-benzodioxole 97%
Catalog # APOSOR9297-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.62 g/mol
Boiling Pt:112 °C (2 mm Hg)
Storage Temperature:Ambient
UN:1760
ADR:8,III
CAS Number:23808-46-0
Formula:C₉H₉ClO₂
MDL Number:MFCD08435910