7-Methoxy-1-methyl-2-tetralone 95%
Catalog # APOSOR929481-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.238 g/mol
Storage Temperature:Ambient
CAS Number:1204-23-5
Formula:C₁₂H₁₄O₂
MDL Number:MFCD00797848