Order Entry
Czech Republic
ContactUsLinkComponent
2-(T-Butyloxy)-ethylamine hydrochloride 95%
2-(T-Butyloxy)-ethylamine hydrochloride 95%
Catalog # APOSOR929232-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(T-Butyloxy)-ethylamine hydrochloride 95%
Catalog # APOSOR929232-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.65 g/mol
Storage Temperature:Ambient
CAS Number:335598-67-9
Formula:C₆H₁₅NO·ClH
MDL Number:MFCD04973191