Order Entry
Czech Republic
ContactUsLinkComponent
5-Formyl-2-isoproxypyridine 95%
5-Formyl-2-isoproxypyridine 95%
Catalog # APOSOR927240-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Formyl-2-isoproxypyridine 95%
Catalog # APOSOR927240-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :165.189 g/mol
Storage Temperature:Ambient
CAS Number:884495-35-6
Formula:C₉H₁₁NO₂
MDL Number:MFCD07368872