
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000341-75-2
:
Formula:C₈H₅BrN₂O
MDL Number:MFCD09749988
:225.042 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together