Order Entry
Czech Republic
ContactUsLinkComponent
1-(Ethylamino)-3-(dimethylamino)propane 95%
1-(Ethylamino)-3-(dimethylamino)propane 95%
Catalog # APOSOR927198-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Ethylamino)-3-(dimethylamino)propane 95%
Catalog # APOSOR927198-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :130.231 g/mol
Storage Temperature:Ambient
CAS Number:19475-27-5
Formula:C₇H₁₈N₂
MDL Number:MFCD00051552