4-[(Methylamino)methyl]phenol 95%
Catalog # APOSOR927144-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.179 g/mol
Storage Temperature:Ambient
CAS Number:78507-19-4
Formula:C₈H₁₁NO
MDL Number:MFCD09046544