Order Entry
Czech Republic
ContactUsLinkComponent
6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol 95%
6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol 95%
Catalog # APOSOR926886-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dihydro-5h-cyclopenta[c]pyridazin-3-ol 95%
Catalog # APOSOR926886-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :136.151 g/mol
Storage Temperature:Ambient
CAS Number:122001-78-9
 :MFCD07757617
Formula:C₇H₈N₂O
MDL Number:MFCD20502855