
5-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 97+%
Catalog # APOSOR926154-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:241.042 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1026201-52-4
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD11043994