
[5-(Fur-2-yl)-1-methyl-1H-pyrazol-3-yl]methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:876728-41-5
:
Formula:C₉H₁₀N₂O₂
MDL Number:MFCD08271948
:178.188 g/mol
Melting Pt:43…48 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together