Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate 95%
tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate 95%
Catalog # APOSOR924245-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl N-[(1R,4S)-4-hydroxycyclopent-2-en-1-yl]carbamate 95%
Catalog # APOSOR924245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.247 g/mol
Storage Temperature:Ambient
CAS Number:178152-48-2
Formula:C₁₀H₁₇NO₃
MDL Number:MFCD09954747