Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylcyclohexane-1,3-dione
2,2-Dimethylcyclohexane-1,3-dione 95%
2,2-Dimethylcyclohexane-1,3-dione 95%
APOSOR923906-1G
:
Apollo Scientific
:
2,2-Dimethylcyclohexane-1,3-dione 95%
APOSOR923906-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
140.18 g/mol
:
Ambient
:
562-13-0
:
C₈H₁₂O₂
:
MFCD14708261