Order Entry
Czech Republic
ContactUsLinkComponent
5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde 97%
Irritant
5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde 97%
Catalog # APOSOR9237-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Fur-2-yl)-1-methyl-1H-pyrazole-3-carboxaldehyde 97%
Catalog # APOSOR9237-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:62…64 °C
 :176.172 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:876728-40-4
Formula:C₉H₈N₂O₂
MDL Number:MFCD08271947