Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate 95%
tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate 95%
Catalog # APOSOR922315-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate 95%
Catalog # APOSOR922315-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.274 g/mol
Storage Temperature:Ambient
CAS Number:1211531-16-6
Formula:C₁₁H₁₉NO₂
MDL Number:MFCD19703948