Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dioxaspiro[4.5]decan-7-one 95%
1,4-Dioxaspiro[4.5]decan-7-one 95%
Catalog # APOSOR922251-1G
Supplier:  Apollo Scientific
CAS Number:  
1,4-Dioxaspiro[4.5]decan-7-one 95%
Catalog # APOSOR922251-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.179 g/mol
Storage Temperature:Ambient
CAS Number:4969-01-1
Formula:C₈H₁₂O₃
MDL Number:MFCD16039576